3-Bromo-4-chloro-5-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
PC303563-1G PC303563-5G PC303563-25G
APOSPC303563-1GEA 219 EUR
APOSPC303563-1G APOSPC303563-5G APOSPC303563-25G
3-Bromo-4-chloro-5-(trifluoromethyl)benzaldehyde 98%
3-Bromo-4-chloro-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₃BrClF₃O
MW: 287.461 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640141
CAS Number: 2385966-70-9