3-Bromo-4-chloro-5-(trifluoromethyl)benzyl bromide
Supplier: Apollo Scientific
PC303565-1G PC303565-5G
APOSPC303565-1GEA 279 EUR
APOSPC303565-1G APOSPC303565-5G
3-Bromo-4-chloro-5-(trifluoromethyl)benzyl bromide
3-Bromo-4-chloro-5-(trifluoromethyl)benzyl bromide
Formula: C₈H₄Br₂ClF₃
MW: 352.374 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640143
CAS Number: 2386636-01-5