3-Bromo-4-chloro-5-(trifluoromethyl)benzoic acid 98%
Supplier: Apollo Scientific
PC303569-1G PC303569-5G
APOSPC303569-1GEA 198 EUR
APOSPC303569-1G APOSPC303569-5G
3-Bromo-4-chloro-5-(trifluoromethyl)benzoic acid 98%
3-Bromo-4-chloro-5-(trifluoromethyl)benzoic acid
Formula: C₈H₃BrClF₃O₂
MW: 303.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640147
CAS Number: 2386483-96-9