5-Bromo-2-methyl-3-(trifluoromethyl)benzyl bromide 97%
Supplier: Apollo Scientific
PC303588-1G PC303588-5G
APOSPC303588-1GEA 348 EUR
APOSPC303588-1G APOSPC303588-5G
5-Bromo-2-methyl-3-(trifluoromethyl)benzyl bromide 97%
5-Bromo-2-methyl-3-(trifluoromethyl)benzyl bromide
Formula: C₉H₇Br₂F₃
MW: 331.955 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640163
CAS Number: 2386352-03-8
UN: 3265
ADR: 8 III