3-Bromo-4-methyl-5-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
PC303599-1G PC303599-5G
APOSPC303599-1GEA 307 EUR
APOSPC303599-1G APOSPC303599-5G
3-Bromo-4-methyl-5-(trifluoromethyl)benzaldehyde 97%
3-Bromo-4-methyl-5-(trifluoromethyl)benzaldehyde
Formula: C₉H₆BrF₃O
MW: 267.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640172
CAS Number: 2383384-91-4