2-Bromo-5-(pentafluorosulphur)aniline 98%
Supplier: Apollo Scientific
PC303612-10G PC303612-1G PC303612-5G
APOSPC303612-10GEA 2490 EUR
APOSPC303612-10G APOSPC303612-1G APOSPC303612-5G
2-Bromo-5-(pentafluorosulphur)aniline 98%
2-Bromo-5-(pentafluorosulphur)aniline
Formula: C₆H₅BrF₅NS
MW: 298.072 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28385432
CAS Number: 1211537-51-7