2-Methyl-3-(trifluoromethyl)-α-methylstyrene 95%
Supplier: Apollo Scientific
PC303614-1G PC303614-250MG PC303614-5G
APOSPC303614-1GEA 522 EUR
APOSPC303614-1G APOSPC303614-250MG APOSPC303614-5G
2-Methyl-3-(trifluoromethyl)-α-methylstyrene 95%
2-Methyl-3-(trifluoromethyl)-α-methylstyrene
Formula: C₁₁H₁₁F₃
MW: 200.2 g/mol
Storage Temperature: Refrigerator
CAS Number: 2514942-04-0