3,3,3-Trifluoropropane-1,2-diamine hydrochloride
Supplier: Apollo Scientific
Warning
PC305002-500MG PC305002-50MG
APOSPC305002-500MGEA 1120 EUR
APOSPC305002-500MG APOSPC305002-50MG
3,3,3-Trifluoropropane-1,2-diamine hydrochloride
3,3,3-Trifluoropropane-1,2-diamine hydrochloride
Formula: C₃H₇F₃N₂·ClH
MW: 164.557 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103316
CAS Number: 928779-95-7