2-(Trifluoromethyl)morpholine hydrochloride
Supplier: Apollo Scientific
Warning
PC305012-1G PC305012-250MG
APOSPC305012-1GEA 607 EUR
APOSPC305012-1G APOSPC305012-250MG
2-(Trifluoromethyl)morpholine hydrochloride
2-(Trifluoromethyl)morpholine hydrochloride
Formula: C₅H₈F₃NO·ClH
Storage Temperature: Ambient
MDL Number: MFCD13189488
CAS Number: 1196152-51-8