6-Bromo-5-fluoronicotinic acid 97%
Supplier: Apollo Scientific
Warning
PC305017-1G PC305017-250MG PC305017-5G
APOSPC305017-1GEA 261 EUR
APOSPC305017-1G APOSPC305017-250MG APOSPC305017-5G
6-Bromo-5-fluoronicotinic acid 97%
6-Bromo-5-fluoronicotinic acid
Formula: C₆H₃BrFNO₂
MW: 219.996 g/mol
Melting Pt: 191…196 °C
Storage Temperature: Ambient
MDL Number: MFCD11036153
CAS Number: 38186-87-7