Methyl 2-methyl-6-(trifluoromethyl)nicotinate
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Ethoxycarbonyl)-2-methyl-6-(trifluoromethyl)pyridine
PC31131-1G PC31131-5G PC31131-10G
APOSPC31131-1GEA 87 EUR
APOSPC31131-1G APOSPC31131-5G APOSPC31131-10G
Methyl 2-methyl-6-(trifluoromethyl)nicotinate
Methyl 2-methyl-6-(trifluoromethyl)nicotinate
Formula: C₉H₈F₃NO₂
MW: 219.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01311989
CAS Number: 205582-88-3