2,4-Bis(trifluoromethyl)-DL-phenylalanine 98%
Supplier: Apollo Scientific
PC3114K-1G PC3114K-5G
APOSPC3114K-1GEA 116 EUR
APOSPC3114K-1G APOSPC3114K-5G
2,4-Bis(trifluoromethyl)-DL-phenylalanine 98%
2,4-Bis(trifluoromethyl)-DL-phenylalanine
Formula: C₁₁H₉F₆NO₂
MW: 301.185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061306
CAS Number: 237076-67-4