2-Bromo-4-(trifluoromethoxy)phenol 97%
Supplier: Apollo Scientific
Warning
PC3121-1G PC3121-5G
APOSPC3121-1GEA 21.9 EUR
APOSPC3121-1G APOSPC3121-5G
2-Bromo-4-(trifluoromethoxy)phenol 97%
2-Bromo-4-(trifluoromethoxy)phenol
Formula: C₇H₄BrF₃O₂
MW: 257.005 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07778431
CAS Number: 200956-13-4