3-(1H,1H,7H-Perfluoroheptyloxy)-1,2-propenoxide 95%
Supplier: Apollo Scientific
Synonyms: 1H,1H,2H,3H,3H,5H,5H-Perfluoro(1,2-Epoxy-4-oxaundecane)
PC3132FB-25G PC3132FB-5G
APOSPC3132FB-25GEA 611 EUR
APOSPC3132FB-25G APOSPC3132FB-5G
3-(1H,1H,7H-Perfluoroheptyloxy)-1,2-propenoxide 95%
3-(1H,1H,7H-Perfluoroheptyloxy)-1,2-propenoxide
Formula: C₁₀H₈F₁₂O₂
MW: 388.15 g/mol
Boiling Pt: 97…98 °C (4 mm Hg)
Density: 1.614 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054696
CAS Number: 799-34-8