2-Fluoropyrimidine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-1,3-diazine
PC3138-1G PC3138-5G
APOSPC3138-1GEA 209 EUR
APOSPC3138-1G APOSPC3138-5G
2-Fluoropyrimidine 97%
2-Fluoropyrimidine
Formula: C₄H₃FN₂
MW: 98.078 g/mol
Boiling Pt: 75 °C (20 mm Hg)
Melting Pt: 17…25 °C
Density: 1.5623 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD08059530
CAS Number: 31575-35-6