1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride 97%
Supplier: Apollo Scientific
Danger
PC31422-1G PC31422-250MG
APOSPC31422-1GEA 215 EUR
APOSPC31422-1G APOSPC31422-250MG
1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride 97%
1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride
Formula: C₁₁H₈ClFN₂O
MW: 238.645 g/mol
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD03659694
CAS Number: 423768-49-4
UN: 1759
ADR: 8 III