4-Chloro-2-[(4-chlorophenyl)sulphonyl]-5-(trifluoromethyl)aniline
Supplier: Apollo Scientific
PC31482-1G PC31482-250MG
APOSPC31482-1GEA 98.9 EUR
APOSPC31482-1G APOSPC31482-250MG
4-Chloro-2-[(4-chlorophenyl)sulphonyl]-5-(trifluoromethyl)aniline
4-Chloro-2-[(4-chlorophenyl)sulphonyl]-5-(trifluoromethyl)aniline
Formula: C₁₃H₈Cl₂F₃NO₂S
MW: 370.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00119596
CAS Number: 634187-71-6