1-Methyl-3-thien-2-yl-5-(trifluoromethyl)-1H-pyrazole
Supplier: Apollo Scientific
PC31812-1G
PC31812-5G
APOSPC31812-1GEA
114
EUR
APOSPC31812-1G
APOSPC31812-5G
1-Methyl-3-thien-2-yl-5-(trifluoromethyl)-1H-pyrazole
1-Methyl-3-thien-2-yl-5-(trifluoromethyl)-1H-pyrazole
Formula:
C₉H₇F₃N₂S MW: 232.225 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02089760 CAS Number: 650615-64-8 |