4-Chloro-6-methyl-2-phenyl-5-[3-(trifluoromethyl)phenoxy]pyrimidine
Supplier: Apollo Scientific
PC32678-1G PC32678-250MG
APOSPC32678-1GEA 98.9 EUR
APOSPC32678-1G APOSPC32678-250MG
4-Chloro-6-methyl-2-phenyl-5-[3-(trifluoromethyl)phenoxy]pyrimidine
4-Chloro-6-methyl-2-phenyl-5-[3-(trifluoromethyl)phenoxy]pyrimidine
Formula: C₁₈H₁₂ClF₃N₂O
MW: 364.749 g/mol
Melting Pt: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00833315
CAS Number: 680217-69-0