Ethyl 5-cyano-6-hydroxy-2-(trifluoromethyl)nicotinate
Supplier: Apollo Scientific
Warning
PC3269-1G PC3269-5G
APOSPC3269-1GEA 102 EUR
APOSPC3269-1G APOSPC3269-5G
Ethyl 5-cyano-6-hydroxy-2-(trifluoromethyl)nicotinate
Ethyl 5-cyano-6-hydroxy-2-(trifluoromethyl)nicotinate
Formula: C₁₀H₇F₃N₂O₃
MW: 260.169 g/mol
Melting Pt: 201…203 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00178796
CAS Number: 144456-86-0