N1-Methyl-4-(trifluoromethyl)benzene-1,2-diamine
Supplier: Apollo Scientific
Danger
Synonyms: 2-Amino-1-(methylamino)-4-(trifluoromethyl)benzene, 3-Amino-4-(methylamino)benzotrifluoride
PC32696-1G PC32696-5G
APOSPC32696-1GEA 154 EUR
APOSPC32696-1G APOSPC32696-5G
N1-Methyl-4-(trifluoromethyl)benzene-1,2-diamine
N1-Methyl-4-(trifluoromethyl)benzene-1,2-diamine
Formula: C₈H₉F₃N₂
MW: 190.166 g/mol
Melting Pt: 53…55 °C
Storage Temperature: Ambient
MDL Number: MFCD01571385
CAS Number: 35203-49-7
UN: 2811
ADR: 6.1 III