2-Bromo-2,2-difluoro-N-(2-hydroxyphenyl)acetamide
Supplier: Apollo Scientific
Warning
PC32711-1G PC32711-5G
APOSPC32711-1GEA 279 EUR
APOSPC32711-1G APOSPC32711-5G
2-Bromo-2,2-difluoro-N-(2-hydroxyphenyl)acetamide
2-Bromo-2,2-difluoro-N-(2-hydroxyphenyl)acetamide
Formula: C₈H₆BrF₂NO₂
MW: 266.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11035865
CAS Number: 239136-81-3