2,4-Bis(trifluoroacetyl)-1-(N,N-dimethylamino)naphthalene
Supplier: Apollo Scientific
Warning
Synonyms: 2,4-Bis(trifluoroacetyl)-N,N-dimethylnaphth-1ylamine
PC32717-1G PC32717-5G
APOSPC32717-1GEA 91.5 EUR
APOSPC32717-1G APOSPC32717-5G
2,4-Bis(trifluoroacetyl)-1-(N,N-dimethylamino)naphthalene
2,4-Bis(trifluoroacetyl)-1-(N,N-dimethylamino)naphthalene
Formula: C₁₆H₁₁F₆NO₂
MW: 363.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11035868
CAS Number: 115975-33-2