(2R)-(+)-3,3,3-Trifluoro-1,2-propenoxide
Supplier: Apollo Scientific
Synonyms: (2R)-1,2-Epoxy-3,3,3-trifluoropropane, (2R)-2-(Trifluoromethyl)oxirane
PC32742-1G PC32742-5G PC32742-250MG
APOSPC32742-1GEA 319 EUR
APOSPC32742-1G APOSPC32742-5G APOSPC32742-250MG
(2R)-(+)-3,3,3-Trifluoro-1,2-propenoxide
(2R)-(+)-3,3,3-Trifluoro-1,2-propenoxide
Formula: C₃H₃F₃O
MW: 112.051 g/mol
Boiling Pt: 25…32 °C
Melting Pt: –80 °C
Density: 1.294 g/cm³ (25 °C)
Flash Pt: –26 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08276876
CAS Number: 143142-90-9
UN: 1993
ADR: 3,I