3-Fluoro-5-iodobenzyl alcohol
Supplier: Apollo Scientific
Warning
Synonyms: (3-Fluoro-5-iodophenyl)methanol
PC32750-1G PC32750-250MG PC32750-5G
APOSPC32750-1GEA 81.2 EUR
APOSPC32750-1G APOSPC32750-250MG APOSPC32750-5G
3-Fluoro-5-iodobenzyl alcohol
3-Fluoro-5-iodobenzyl alcohol
Formula: C₇H₆FIO
MW: 252.025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18396948
CAS Number: 1261837-87-9