4-Bromo-2,6-difluorobenzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
PC32846-1G PC32846-250MG PC32846-5G
APOSPC32846-1GEA 261 EUR
APOSPC32846-1G APOSPC32846-250MG APOSPC32846-5G
4-Bromo-2,6-difluorobenzenesulphonyl chloride
4-Bromo-2,6-difluorobenzenesulphonyl chloride
Formula: C₆H₂BrClF₂O₂S
MW: 291.498 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06409102
CAS Number: 874804-21-4
UN: 3261
ADR: 8,III