2,6-Difluoro-N-hydroxybenzamide
Supplier: Apollo Scientific
PC32871-1G PC32871-250MG PC32871-500MG PC32871-2.5G
APOSPC32871-1GEA 484 EUR
APOSPC32871-1G APOSPC32871-250MG APOSPC32871-500MG APOSPC32871-2.5G
2,6-Difluoro-N-hydroxybenzamide
2,6-Difluoro-N-hydroxybenzamide
Formula: C₇H₅F₂NO₂
MW: 173.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03551082
CAS Number: 125309-34-4