Ethyl 4-fluorobenzofuran-2-carboxylate
Supplier: Apollo Scientific
Warning
PC32888-1G PC32888-250MG
APOSPC32888-1GEA 178 EUR
APOSPC32888-1G APOSPC32888-250MG
Ethyl 4-fluorobenzofuran-2-carboxylate
Ethyl 4-fluorobenzofuran-2-carboxylate
Formula: C₁₁H₉FO₃
MW: 208.186 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30528090
CAS Number: 1709824-79-2