4-Fluoro-1-benzofuran-2-carboxylic acid
Supplier: Apollo Scientific
Warning
Synonyms: 4-Fluoro-benzofuran-2-carboxylic acid
PC32889-1G PC32889-250MG
APOSPC32889-1GEA 493 EUR
APOSPC32889-1G APOSPC32889-250MG
4-Fluoro-1-benzofuran-2-carboxylic acid
4-Fluoro-1-benzofuran-2-carboxylic acid
Formula: C₉H₅FO₃
MW: 180.133 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23708314
CAS Number: 1408060-30-9