2-Oxo-2-(3-(trifluoromethyl)phenyl)acetic acid 97%
Supplier: Apollo Scientific
Warning
PC32903-1G PC32903-250MG
APOSPC32903-1GEA 139 EUR
APOSPC32903-1G APOSPC32903-250MG
2-Oxo-2-(3-(trifluoromethyl)phenyl)acetic acid 97%
2-Oxo-2-(3-(trifluoromethyl)phenyl)acetic acid
Formula: C₉H₅F₃O₃
MW: 218.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07777206
CAS Number: 61560-95-0