2-(Carboxymethyl)-6-fluorobenzoic acid 97%
Supplier: Apollo Scientific
Warning
PC32907-1G PC32907-250MG
APOSPC32907-1GEA 522 EUR
APOSPC32907-1G APOSPC32907-250MG
2-(Carboxymethyl)-6-fluorobenzoic acid 97%
2-(Carboxymethyl)-6-fluorobenzoic acid
Formula: C₉H₇FO₄
MW: 198.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20639259
CAS Number: 583881-07-6