2-[Bromo(difluoro)methyl]-4-(trifluoromethoxy)-1H-benzimidazole
Supplier: Apollo Scientific
Warning
PC32925-1G PC32925-250MG PC32925-5G
APOSPC32925-1GEA 63.8 EUR
APOSPC32925-1G APOSPC32925-250MG APOSPC32925-5G
2-[Bromo(difluoro)methyl]-4-(trifluoromethoxy)-1H-benzimidazole
2-[Bromo(difluoro)methyl]-4-(trifluoromethoxy)-1H-benzimidazole
Formula: C₉H₄BrF₅N₂O
MW: 331.037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31922508
CAS Number: 2366994-28-5