4-Fluoro-5-iodo-1H-pyrrolo[2,3-b]pyridine 99%
Supplier: Apollo Scientific
Warning
PC32958-1G PC32958-250MG
APOSPC32958-1GEA 427 EUR
APOSPC32958-1G APOSPC32958-250MG
4-Fluoro-5-iodo-1H-pyrrolo[2,3-b]pyridine 99%
4-Fluoro-5-iodo-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₄FIN₂
MW: 262.023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986882
CAS Number: 1638765-44-2