1,1,1,3,3,3-Hexafluoro-2-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-ol 95%
Supplier: Apollo Scientific
Warning
PC32964-1G PC32964-250MG
APOSPC32964-1GEA 69.6 EUR
APOSPC32964-1G APOSPC32964-250MG
1,1,1,3,3,3-Hexafluoro-2-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-ol 95%
1,1,1,3,3,3-Hexafluoro-2-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-ol
Formula: C₁₅H₁₆BF₇O₃
MW: 388.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32070723
CAS Number: 2376598-54-6