2-(Difluoromethyl)-1H-benzimidazol-6-amine
Supplier: Apollo Scientific
Warning
PC32966-1G PC32966-250MG
APOSPC32966-1GEA 250 EUR
APOSPC32966-1G APOSPC32966-250MG
2-(Difluoromethyl)-1H-benzimidazol-6-amine
2-(Difluoromethyl)-1H-benzimidazol-6-amine
Formula: C₈H₇F₂N₃
MW: 183.158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28665942
CAS Number: 891831-55-3