1-(3,5-Difluorophenyl)-imidazolidin-2-one
Supplier: Apollo Scientific
Warning
PC32978-1G PC32978-250MG
APOSPC32978-1GEA 364 EUR
APOSPC32978-1G APOSPC32978-250MG
1-(3,5-Difluorophenyl)-imidazolidin-2-one
1-(3,5-Difluorophenyl)-imidazolidin-2-one
Formula: C₉H₈F₂N₂O
MW: 198.169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23696670
CAS Number: 1547051-49-9