1-(2,4-Difluorophenyl)-imidazolidin-2-one
Supplier: Apollo Scientific
Warning
PC32979-1G PC32979-250MG
APOSPC32979-1GEA 84.1 EUR
APOSPC32979-1G APOSPC32979-250MG
1-(2,4-Difluorophenyl)-imidazolidin-2-one
1-(2,4-Difluorophenyl)-imidazolidin-2-one
Formula: C₉H₈F₂N₂O
MW: 198.169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24189713
CAS Number: 1545294-47-0