1-(3-Trifluoromethoxyphenyl)-imidazolidin-2-one
Supplier: Apollo Scientific
Warning
PC32981-1G PC32981-250MG
APOSPC32981-1GEA 102 EUR
APOSPC32981-1G APOSPC32981-250MG
1-(3-Trifluoromethoxyphenyl)-imidazolidin-2-one
1-(3-Trifluoromethoxyphenyl)-imidazolidin-2-one
Formula: C₁₀H₉F₃N₂O₂
MW: 246.186 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23696675
CAS Number: 1554285-69-6