1H,1H,2H-Pentacosafluorotetradec-1-ene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-Pentacosafluorotetradec-1-ene, (Perfluorododec-1-yl)ethylene
PC3337-1G PC3337-5G
APOSPC3337-1GEA 98.9 EUR
APOSPC3337-1G APOSPC3337-5G
1H,1H,2H-Pentacosafluorotetradec-1-ene 97%
1H,1H,2H-Pentacosafluorotetradec-1-ene
Formula: C₁₄H₃F₂₅
MW: 646.134 g/mol
Boiling Pt: 118…120 °C (20 mm Hg)
Melting Pt: 61…62 °C
Storage Temperature: Ambient
MDL Number: MFCD04038351
CAS Number: 67103-05-3