2,2,2-Trifluoro-1-(trifluoromethyl)ethylamine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1,1,1,3,3,3-Hexafluoropropan-2-amine, 2-Amino-2H-hexafluoropropane
PC3608-1G PC3608-5G
APOSPC3608-1GEA 157 EUR
APOSPC3608-1G APOSPC3608-5G
2,2,2-Trifluoro-1-(trifluoromethyl)ethylamine 97%
2,2,2-Trifluoro-1-(trifluoromethyl)ethylamine
Formula: C₃H₃F₆N
Boiling Pt: 57…58 °C
Density: 1.5442 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD08460498
CAS Number: 1619-92-7
UN: 2920
ADR: 8+3,II