4,4,4-Trifluorobutylamine hydrochloride 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4,4,4-Trifluorobutan-1-amine hydrochloride
PC3636-1G PC3636-250MG PC3636-5G
APOSPC3636-1GEA 139 EUR
APOSPC3636-1G APOSPC3636-250MG APOSPC3636-5G
4,4,4-Trifluorobutylamine hydrochloride 95%
4,4,4-Trifluorobutylamine hydrochloride
Formula: C₄H₈F₃N·ClH
MW: 163.569 g/mol
Melting Pt: 180…186 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD09258951
CAS Number: 84153-82-2