(1R)-1-[4-(Trifluoromethyl)phenyl]ethylamine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-[(1R)-1-Aminoethyl]benzotrifluoride, (R)-1-[4-(Trifluoromethyl)phenyl]ethylamine, (R)-α-Methyl-4-(trifluoromethyl)benzylamine
PC3647-1G PC3647-250MG
APOSPC3647-1GEA 310 EUR
APOSPC3647-1G APOSPC3647-250MG
(1R)-1-[4-(Trifluoromethyl)phenyl]ethylamine 98%
(1R)-1-[4-(Trifluoromethyl)phenyl]ethylamine
Formula: C₉H₁₀F₃N
MW: 189.178 g/mol
Boiling Pt: 76 °C (20 mm Hg)
Density: 1.03 g/cm³ (25 °C)
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06761837
CAS Number: 578027-35-7