2-(3,4-Dimethylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Supplier: Apollo Scientific
Warning
Synonyms: 1,2-Dimethyl-4-[perfluoro(2-hydroxyprop-2-yl)]benzene, 4-(1,1,1,3,3,3-Hexafluoro-2-hydroxyprop-2-yl)-o-xylene
PC3741-1G PC3741-25G PC3741-5G
APOSPC3741-1GEA 58 EUR
APOSPC3741-1G APOSPC3741-25G APOSPC3741-5G
2-(3,4-Dimethylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
2-(3,4-Dimethylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Formula: C₁₁H₁₀F₆O
MW: 272.187 g/mol
Boiling Pt: 101…102 °C (20 mm Hg)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03412228
CAS Number: 2379-17-1