Ethyl 6-cyano-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Supplier: Apollo Scientific
Warning
PC37865-1G PC37865-250MG
APOSPC37865-1GEA 522 EUR
APOSPC37865-1G APOSPC37865-250MG
Ethyl 6-cyano-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Ethyl 6-cyano-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Formula: C₁₂H₈F₃N₃O₂
MW: 283.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32200427
CAS Number: 2407339-54-0