2-Bromo-1-(6-bromo-2,3,4-trifluorophenyl)ethanone 95%
Supplier: Apollo Scientific
PC38225-1G PC38225-250MG
APOSPC38225-1GEA 654 EUR
APOSPC38225-1G APOSPC38225-250MG
2-Bromo-1-(6-bromo-2,3,4-trifluorophenyl)ethanone 95%
2-Bromo-1-(6-bromo-2,3,4-trifluorophenyl)ethanone
Formula: C₈H₃Br₂F₃O
MW: 331.912 g/mol
Storage Temperature: Ambient
CAS Number: 2416368-70-0
UN: 3261
ADR: 8,III