3-Fluoro-4-(methylthio)benzamide 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Fluoro-4-(methylsulphanyl)benzamide, 4-Carbamoyl-2-fluorothioanisole
PC3829-1G PC3829-250MG
APOSPC3829-1GEA 621 EUR
APOSPC3829-1G APOSPC3829-250MG
3-Fluoro-4-(methylthio)benzamide 97%
3-Fluoro-4-(methylthio)benzamide
Formula: C₈H₈FNOS
MW: 185.219 g/mol
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00673179
CAS Number: 238742-81-9