2,6-Difluorostyrene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1,3-Difluoro-2-ethenylbenzene, 1,3-Difluoro-2-vinylbenzene
PC3951-1G PC3951-5G
APOSPC3951-1GEA 43.5 EUR
APOSPC3951-1G APOSPC3951-5G
2,6-Difluorostyrene 97%
2,6-Difluorostyrene
Formula: C₈H₆F₂
MW: 140.13 g/mol
Boiling Pt: 59 °C (51 mm Hg)
Density: 1.13 g/cm³ (25 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00134389
CAS Number: 207226-37-7
UN: 1993
ADR: 3,III