2-(2,2-Dibromovinyl)-4-chloro-1-fluorobenzene
Supplier: Apollo Scientific
Danger
PC400043-250MG PC400043-1G
APOSPC400043-250MGEA 154 EUR
APOSPC400043-250MG APOSPC400043-1G
2-(2,2-Dibromovinyl)-4-chloro-1-fluorobenzene
2-(2,2-Dibromovinyl)-4-chloro-1-fluorobenzene
Formula: C₈H₄Br₂ClF
MW: 314.377 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561229
CAS Number: 1881272-50-9
UN: 2810
ADR: 6.1,III