1-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)-2,2,2-trifluoroethanone
Supplier: Apollo Scientific
Warning
PC400148-1G PC400148-250MG
APOSPC400148-1GEA 569 EUR
APOSPC400148-1G APOSPC400148-250MG
1-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)-2,2,2-trifluoroethanone
1-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)-2,2,2-trifluoroethanone
Formula: C₈H₂ClF₆NO
MW: 277.551 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31621152
CAS Number: 2231675-30-0