6-(Trifluoromethyl)-[1,2,4]triazolo[1,5-a]pyridin-2-amine
Supplier: Apollo Scientific
Warning
PC400172-1G PC400172-250MG
APOSPC400172-1GEA 120 EUR
APOSPC400172-1G APOSPC400172-250MG
6-(Trifluoromethyl)-[1,2,4]triazolo[1,5-a]pyridin-2-amine
6-(Trifluoromethyl)-[1,2,4]triazolo[1,5-a]pyridin-2-amine
Formula: C₇H₅F₃N₄
MW: 202.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847748
CAS Number: 1239648-22-6